Product Details

SNP ID
rs181739156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:64979407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTGCTGTTTACACTCAACCACA[C/G]TTTTACAGCCTTTTCTTAACTGTAA
Phenotype
MIM: 194624
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF117 PubMed Links

Gene Details

Gene
ZNF117
Gene Name
zinc finger protein 117
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015852.3 1449 Missense Mutation ACT,AGT T55S NP_056936.2

View Full Product Details