Product Details

SNP ID
rs181707288
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:200891599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGGAGGGCAGGCCGCAGGGAGG[C/T]TCGGGGGGAGGGAAGATGGCCTGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf106 PubMed Links

Gene Details

Gene
C1orf106
Gene Name
chromosome 1 open reading frame 106
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142569.2 101 Intron NP_001136041.1
NM_018265.3 101 Missense Mutation GCT,GTT A34V NP_060735.3
XM_011509754.2 101 Intron XP_011508056.1
XM_011509755.1 101 Intron XP_011508057.1
XM_011509756.2 101 Missense Mutation GCT,GTT A34V XP_011508058.1

View Full Product Details