Product Details

SNP ID
rs181498009
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:149905921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTTCAACAGTCAACACGTCCAG[C/T]GCATTCCAGGATGCAATGCTGACCC
Phenotype
MIM: 613181 MIM: 185860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BOLA1 PubMed Links

Gene Details

Gene
BOLA1
Gene Name
bolA family member 1
There are no transcripts associated with this gene.

Gene
SV2A
Gene Name
synaptic vesicle glycoprotein 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278719.1 2281 Silent Mutation GCA,GCG A120A NP_001265648.1
NM_014849.4 2281 Silent Mutation GCA,GCG A668A NP_055664.3
XM_017002992.1 2281 Silent Mutation GCA,GCG A668A XP_016858481.1

View Full Product Details