Product Details

SNP ID
rs181568602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49425423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGGTGGGAGAGCCACTTGGGAA[A/G]GGTCACGGAGGGGCTCTGCAAAGCA
Phenotype
MIM: 238310 MIM: 611516 MIM: 600690
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMT PubMed Links

Gene Details

Gene
AMT
Gene Name
aminomethyltransferase
There are no transcripts associated with this gene.

Gene
NICN1
Gene Name
nicolin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032316.3 524 Missense Mutation CTT,TTT L147F NP_115692.1
Gene
TCTA
Gene Name
T-cell leukemia translocation altered
There are no transcripts associated with this gene.

View Full Product Details