Product Details

SNP ID
rs182139935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85345997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTGGCAGGCAGCGGGCGTTCCC[C/T]GGCAGTAGGTGTTCATAGGTGTTGA
Phenotype
MIM: 615427
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELMOD3 PubMed Links

Gene Details

Gene
ELMOD3
Gene Name
ELMO domain containing 3
There are no transcripts associated with this gene.

Gene
RETSAT
Gene Name
retinol saturase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017750.3 1286 Silent Mutation CCA,CCG P365P NP_060220.3

View Full Product Details