Product Details

SNP ID
rs182187602
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:152719740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCTACGTGAAGTGCCCAGCTCC[C/T]TGCCAGAGGACCTATGTGAAATACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf68 PubMed Links

Gene Details

Gene
C1orf68
Gene Name
chromosome 1 open reading frame 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024679.2 219 Silent Mutation CCC,CCT P73P NP_001019850.1

View Full Product Details