Product Details

SNP ID
rs182322273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76390319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACAGTGCCCCACCGTTGCCCTA[C/G]GTAGAGCTCTACACGTCCCTCGCAT
Phenotype
MIM: 607639 MIM: 182889
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SSC4D PubMed Links

Gene Details

Gene
SSC4D
Gene Name
scavenger receptor cysteine rich family member with 4 domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080744.1 1605 Missense Mutation CTA,GTA L490V NP_542782.1
XM_017011750.1 1605 Missense Mutation CTA,GTA L321V XP_016867239.1
Gene
ZP3
Gene Name
zona pellucida glycoprotein 3 (sperm receptor)
There are no transcripts associated with this gene.

View Full Product Details