Product Details

SNP ID
rs181996868
Assay Type
Functionally Tested
NCBI dbSNP Submissions
13
Location
Chr.1:37540515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCACCAACGTCATTCCTCTGGC[A/G]ATGCTCCCGTCGCCTGGCATTATAA
Phenotype
MIM: 608241
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SNIP1 PubMed Links

Gene Details

Gene
SNIP1
Gene Name
Smad nuclear interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024700.3 683 Missense Mutation CGC,TGC R190C NP_078976.2

View Full Product Details