Product Details

SNP ID
rs181993007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109689507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGGAGGAGCCATATTTTCCCAA[A/C]ATCTCACACAAATCTTGAATGTACC
Phenotype
MIM: 601532
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CASP6 PubMed Links

Gene Details

Gene
CASP6
Gene Name
caspase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001226.3 516 Missense Mutation ATG,ATT M235I NP_001217.2
NM_032992.2 516 Missense Mutation ATG,ATT M146I NP_116787.1
Gene
MCUB
Gene Name
mitochondrial calcium uniporter dominant negative beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017918.4 516 Intron NP_060388.2
XM_006714246.3 516 Intron XP_006714309.1

View Full Product Details