Product Details

SNP ID
rs182131786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:67397607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCGGGGCTAGAGAGGGGCCTCC[A/C]GGGCCAGTGCGGCAGCGAGAGCAGC
Phenotype
MIM: 613196
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ETAA1 PubMed Links

Gene Details

Gene
ETAA1
Gene Name
Ewing tumor associated antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019002.3 291 Silent Mutation CCA,CCC P53P NP_061875.2
XM_017004376.1 291 Silent Mutation CCA,CCC P53P XP_016859865.1
XM_017004377.1 291 Silent Mutation CCA,CCC P53P XP_016859866.1

View Full Product Details