Product Details

SNP ID
rs181836391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:60760140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTATTTTCTTGAACAGAAACAC[C/T]GTGCTGGACAGCCAGCGTCAACAAA
Phenotype
MIM: 616865
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAPOLG PubMed Links

Gene Details

Gene
PAPOLG
Gene Name
poly(A) polymerase gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022894.3 247 Silent Mutation ACC,ACT T8T NP_075045.2
XM_005264500.3 247 Silent Mutation ACC,ACT T8T XP_005264557.1
XM_005264501.1 247 UTR 5 XP_005264558.1
XM_011533041.1 247 UTR 5 XP_011531343.1
XM_017004723.1 247 UTR 5 XP_016860212.1
XM_017004724.1 247 UTR 5 XP_016860213.1

View Full Product Details