Product Details

SNP ID
rs182459489
Assay Type
Functionally Tested
NCBI dbSNP Submissions
13
Location
Chr.1:223394271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGATTGTGGCCCTGGTGCTGACC[C/T]GTCTCAAGAAGGCCCAGAGGATACG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCDC185 PubMed Links

Gene Details

Gene
CCDC185
Gene Name
coiled-coil domain containing 185
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152610.2 899 Missense Mutation CGT,TGT R266C NP_689823.2

View Full Product Details