Product Details

SNP ID
rs182469194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5985970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTTTGTGTAAACACTCTGAGCA[C/T]TCGCTGGCGGATCTGCTGAGTCTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR52L1 PubMed Links

Gene Details

Gene
OR52L1
Gene Name
olfactory receptor family 52 subfamily L member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005173.3 1016 Missense Mutation ATG,GTG M321V NP_001005173.3

View Full Product Details