Product Details

SNP ID
rs182442940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144004296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTTCACACTCTGTATGATACA[A/T]CTGTACTTCTTCATTGCTCTCATGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR6B1 PubMed Links

Gene Details

Gene
OR6B1
Gene Name
olfactory receptor family 6 subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005281.1 300 Missense Mutation CAA,CAT Q100H NP_001005281.1

View Full Product Details