Product Details

SNP ID
rs182565676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178713267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATCTGCTGTAATTTTTGGTTGA[C/T]TAAGTAATTGTGAATTCTGTTTGAG
Phenotype
MIM: 602444
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724657 PubMed Links

Gene Details

Gene
LOC102724657
Gene Name
uncharacterized LOC102724657
There are no transcripts associated with this gene.

Gene
ZNF354A
Gene Name
zinc finger protein 354A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324339.1 1316 Missense Mutation AAT,AGT N162S NP_001311268.1
NM_005649.2 1316 Missense Mutation AAT,AGT N204S NP_005640.2
XM_011534645.2 1316 Missense Mutation AAT,AGT N162S XP_011532947.1
XM_017009791.1 1316 Missense Mutation AAT,AGT N204S XP_016865280.1

View Full Product Details