Product Details
- SNP ID
-
rs182610822
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:94021508 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACTATGTACCCCAAGGGAAGAATG[C/T]TATTACTGGGAAAAGTTTCACAGAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HEPHL1
PubMed Links
Gene Details
- Gene
- HEPHL1
- Gene Name
- hephaestin like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001098672.1 |
297 |
Missense Mutation |
GCT,GTT |
A47V |
NP_001092142.1 |
View Full Product Details