Product Details

SNP ID
rs182345703
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:119836137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGATTGGGACACATCATGGTCA[A/C]TACAAGAAGTCGTAACCCTTTCCTC
Phenotype
MIM: 613997
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NBPF7 PubMed Links

Gene Details

Gene
NBPF7
Gene Name
neuroblastoma breakpoint family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001047980.2 1331 Missense Mutation AGT,ATT S329I NP_001041445.1
XM_017001186.1 1331 Intron XP_016856675.1

View Full Product Details