Product Details

SNP ID
rs182351306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:174241183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTGCTATTTCCTTTGGCGAATC[A/T]TCCAGGATACCCTTGGACATAAAGT
Phenotype
MIM: 605649
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXO8 PubMed Links

Gene Details

Gene
FBXO8
Gene Name
F-box protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012180.2 1342 Missense Mutation GAA,GAT E164D NP_036312.2
XM_017008020.1 1342 Missense Mutation GAA,GAT E17D XP_016863509.1

View Full Product Details