Product Details

SNP ID
rs182401295
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:45382569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGCGCAAGTACTGGCTGAATGA[C/T]GACTGGTACCTGAAGTACATCACGC
Phenotype
MIM: 152390
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ALOX5 PubMed Links

Gene Details

Gene
ALOX5
Gene Name
arachidonate 5-lipoxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000698.4 351 Silent Mutation GAC,GAT D79D NP_000689.1
NM_001256153.2 351 Silent Mutation GAC,GAT D79D NP_001243082.1
NM_001256154.2 351 Silent Mutation GAC,GAT D79D NP_001243083.1
NM_001320861.1 351 Silent Mutation GAC,GAT D79D NP_001307790.1
NM_001320862.1 351 UTR 5 NP_001307791.1
XM_017016012.1 351 UTR 5 XP_016871501.1

View Full Product Details