Product Details

SNP ID
rs182999910
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:56496667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTAGTCTTGAAGAGGTCAGACA[C/T]GAAGAAAACCTAGAAGCACAAATCA
Phenotype
MIM: 607125
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLPP3 PubMed Links

Gene Details

Gene
PLPP3
Gene Name
phospholipid phosphatase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003713.4 1388 Missense Mutation ATG,GTG M274V NP_003704.3

View Full Product Details