Product Details

SNP ID
rs879019512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:60768500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCTTCTGTTGTGGCTACTGTT[G/T]GTGGTAAAATTTTCACATTTGGATC
Phenotype
MIM: 616865
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PAPOLG PubMed Links

Gene Details

Gene
PAPOLG
Gene Name
poly(A) polymerase gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022894.3 526 Missense Mutation GGT,TGT G93C NP_075045.2

View Full Product Details