Product Details

SNP ID
rs183071247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74635550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCTAGGTGCCTTCCACAGCTGA[C/T]ACCCTGAGCCCCCTTCACATGAGGT
Phenotype
MIM: 602990 MIM: 609842
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLK3 PubMed Links

Gene Details

Gene
CLK3
Gene Name
CDC like kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130028.1 1324 Intron NP_001123500.1
NM_003992.4 1324 Intron NP_003983.2
XM_005254151.3 1324 Intron XP_005254208.1
XM_011521205.2 1324 Intron XP_011519507.1
XM_011521206.2 1324 Intron XP_011519508.1
XM_011521209.2 1324 Intron XP_011519511.2
XM_017021905.1 1324 Intron XP_016877394.1
XM_017021906.1 1324 Intron XP_016877395.1
XM_017021907.1 1324 Intron XP_016877396.1
XM_017021908.1 1324 Intron XP_016877397.1
XM_017021909.1 1324 Intron XP_016877398.1
XM_017021910.1 1324 Intron XP_016877399.1
Gene
EDC3
Gene Name
enhancer of mRNA decapping 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142443.1 1324 Missense Mutation ATC,GTC I351V NP_001135915.1
NM_001142444.1 1324 Missense Mutation ATC,GTC I351V NP_001135916.1
NM_025083.3 1324 Missense Mutation ATC,GTC I351V NP_079359.2
XM_011522092.2 1324 Missense Mutation ATC,GTC I168V XP_011520394.1

View Full Product Details