Product Details

SNP ID
rs182891031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52433973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTAGGGGCAATTGTCATTCAGC[A/G]ATGTGGCCATAGAATTCTCTCAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF534 PubMed Links

Gene Details

Gene
ZNF534
Gene Name
zinc finger protein 534
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143938.1 95 Missense Mutation AAT,GAT N12D NP_001137410.1
NM_001143939.1 95 Missense Mutation AAT,GAT N12D NP_001137411.1
NM_001291368.1 95 Missense Mutation AAT,GAT N12D NP_001278297.1
NM_001291369.1 95 Missense Mutation AAT,GAT N12D NP_001278298.1

View Full Product Details