Product Details

SNP ID
rs183558146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:51490255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGATAATCATTTTTTAAAACAA[C/T]ATAACCACAGTACCATATCACATCT
Phenotype
MIM: 300528
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01496 PubMed Links

Gene Details

Gene
LINC01496
Gene Name
long intergenic non-protein coding RNA 1496
There are no transcripts associated with this gene.

Gene
NUDT11
Gene Name
nudix hydrolase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018159.3 2152 UTR 3 NP_060629.2

View Full Product Details