Product Details

SNP ID
rs183796688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39515875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACCCGTCGGCCATCGGGCTGGC[C/G]GATCCCCCCATTCCCAGTCCTGTCC
Phenotype
MIM: 602768 MIM: 607919
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DLL3 PubMed Links

Gene Details

Gene
DLL3
Gene Name
delta like canonical Notch ligand 3
There are no transcripts associated with this gene.

Gene
SELV
Gene Name
selenoprotein V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182704.1 763 Silent Mutation GCC,GCG A221A NP_874363.1

View Full Product Details