Product Details

SNP ID
rs183927815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98283044 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTGGAACCTTGGTCTGATTGCT[C/T]TTATCTGGAATGACCCACAACTTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5H2 PubMed Links

Gene Details

Gene
OR5H2
Gene Name
olfactory receptor family 5 subfamily H member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005482.1 157 Missense Mutation CTT,TTT L53F NP_001005482.1

View Full Product Details