Product Details

SNP ID
rs183963526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99547322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAAATATATCATTGTTTTTCCC[C/G]TGCATTTTCAGGCTTAATTCATTAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM200A PubMed Links

Gene Details

Gene
FAM200A
Gene Name
family with sequence similarity 200 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145111.3 1270 Missense Mutation CAC,CAG H362Q NP_659802.1
XM_011515911.2 1270 Missense Mutation CAC,CAG H362Q XP_011514213.1
XM_011515913.2 1270 Missense Mutation CAC,CAG H362Q XP_011514215.1

View Full Product Details