Product Details

SNP ID
rs183939689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20775843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGGTTTTTCTTTCCTCAGGCTG[C/G]AAAGAAACCTTCAAATCCAGCCTTC
Phenotype
MIM: 145505
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACSM3 PubMed Links

Gene Details

Gene
ACSM3
Gene Name
acyl-CoA synthetase medium-chain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005622.3 405 Missense Mutation GCA,GGA A75G NP_005613.2
NM_202000.2 405 Missense Mutation GCA,GGA A75G NP_973729.1
XM_011545911.1 405 Missense Mutation GCA,GGA A75G XP_011544213.1
XM_017023523.1 405 Missense Mutation GCA,GGA A75G XP_016879012.1
XM_017023524.1 405 Missense Mutation GCA,GGA A75G XP_016879013.1
Gene
ERI2
Gene Name
ERI1 exoribonuclease family member 2
There are no transcripts associated with this gene.

View Full Product Details