Product Details

SNP ID
rs183846665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47846275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAAGTCCGTCGGGAACATGCCAG[C/T]TAACGAGGACGCTCCCCAGCCAGGG
Phenotype
MIM: 314998
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF81 PubMed Links

Gene Details

Gene
ZNF81
Gene Name
zinc finger protein 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007137.3 257 Missense Mutation GCT,GTT A3V NP_009068.2
XM_005272600.3 257 Missense Mutation GCT,GTT A3V XP_005272657.1
XM_011543899.2 257 Missense Mutation GCT,GTT A3V XP_011542201.1
XM_011543900.2 257 Missense Mutation GCT,GTT A3V XP_011542202.1
XM_017029486.1 257 Missense Mutation GCT,GTT A3V XP_016884975.1
XM_017029487.1 257 Missense Mutation GCT,GTT A3V XP_016884976.1

View Full Product Details