Product Details
- SNP ID
-
rs183854172
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:20144139 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCGGCCTTTACACTATTCAACCA[A/T]CATGAACCCTAGAGCCTGCTATGCA
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR4N5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74033809] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR4N5
- Gene Name
- olfactory receptor family 4 subfamily N member 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004724.1 |
404 |
Missense Mutation |
AAC,ATC |
N135I |
NP_001004724.1 |
View Full Product Details