Product Details

SNP ID
rs183853877
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:156582297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGTTCCCGCTGGCCGTGGAGG[A/C]CTTCCTGCAGGCCCTGCCCCTGTGC
Phenotype
MIM: 608862
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IQGAP3 PubMed Links

Gene Details

Gene
IQGAP3
Gene Name
IQ motif containing GTPase activating protein 3
There are no transcripts associated with this gene.

Gene
NAXE
Gene Name
NAD(P)HX epimerase
There are no transcripts associated with this gene.

Gene
TTC24
Gene Name
tetratricopeptide repeat domain 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105669.2 809 Missense Mutation GAC,GCC D258A NP_001099139.2
XM_011509253.2 809 Missense Mutation GAC,GCC D55A XP_011507555.1
XM_011509254.2 809 Missense Mutation GAC,GCC D55A XP_011507556.1
XM_011509255.2 809 Missense Mutation GAC,GCC D55A XP_011507557.1
XM_017000497.1 809 Intron XP_016855986.1

View Full Product Details