Product Details

SNP ID
rs183749110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5544977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATATGCTCACAGTATGTGTGGG[G/T]TATGATGCGTGTCCTGCAGAAAGGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR52H1 PubMed Links

Gene Details

Gene
OR52H1
Gene Name
olfactory receptor family 52 subfamily H member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005289.1 547 Missense Mutation ACC,CCC T183P NP_001005289.1

View Full Product Details