Product Details

SNP ID
rs183463652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:170252488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTGGTCGGGTAATATAAGAAAC[A/G]TACCACTCTTCATTTAATGAATTCT
Phenotype
MIM: 601603
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C5orf58 PubMed Links

Gene Details

Gene
C5orf58
Gene Name
chromosome 5 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102609.2 1476 Intron NP_001096079.1
NM_001305393.1 1476 Intron NP_001292322.1
NM_001305394.1 1476 Intron NP_001292323.1
NM_001305395.1 1476 Intron NP_001292324.1
NM_001305396.1 1476 Intron NP_001292325.1
NM_001305397.1 1476 Intron NP_001292326.1
XM_011534432.2 1476 Intron XP_011532734.1
XM_017009029.1 1476 Intron XP_016864518.1
XM_017009030.1 1476 Intron XP_016864519.1
XM_017009031.1 1476 Intron XP_016864520.1
XM_017009032.1 1476 Intron XP_016864521.1
XM_017009033.1 1476 Intron XP_016864522.1
Gene
LCP2
Gene Name
lymphocyte cytosolic protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005565.3 1476 Silent Mutation TAC,TAT Y423Y NP_005556.1

View Full Product Details