Product Details

SNP ID
rs184110624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:24956670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCTTCACCTGGCTGATACAGC[A/T]GCTGTTCCTTTCCGGTTGTGGCGCA
Phenotype
MIM: 182279
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SNRPN PubMed Links

Gene Details

Gene
SNRPN
Gene Name
small nuclear ribonucleoprotein polypeptide N
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003097.4 Intron NP_003088.1
NM_022805.3 Intron NP_073716.1
NM_022806.3 Intron NP_073717.1
NM_022807.3 Intron NP_073718.1
NM_022808.3 Intron NP_073719.1
Gene
SNURF
Gene Name
SNRPN upstream reading frame
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005678.4 Intron NP_005669.2
NM_022804.2 Intron NP_073715.1

View Full Product Details