Product Details

SNP ID
rs184227086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:22209283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCTCACAGTGTAGTAGGTCATC[A/G]TCTGTGCTTTGCTGGCAACTACTTT
Phenotype
MIM: 613249
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EBLN1 PubMed Links

Gene Details

Gene
EBLN1
Gene Name
endogenous Bornavirus-like nucleoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199938.1 701 Missense Mutation ACG,ATG T234M NP_001186867.1

View Full Product Details