Product Details
- SNP ID
-
rs184049283
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:92606367 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACGAACTGGAGGATTGGCTGACAA[A/G]AGCTCAAGGAAAACATACACTTTTG
- Phenotype
-
MIM: 148760
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIF11
PubMed Links
Gene Details
- Gene
- KIF11
- Gene Name
- kinesin family member 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004523.3 |
488 |
Silent Mutation |
AAA,AAG |
K60K |
NP_004514.2 |
View Full Product Details