Product Details

SNP ID
rs184600542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76039783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTCTGTTCCAACTTGTCCTCAA[A/G]GGGTGGGAAAGCCACATGGTTGAGG
Phenotype
MIM: 604858
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SRP68 PubMed Links

Gene Details

Gene
SRP68
Gene Name
signal recognition particle 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260502.1 1842 Missense Mutation CTT,TTT L565F NP_001247431.1
NM_001260503.1 1842 Missense Mutation CTT,TTT L264F NP_001247432.1
NM_014230.3 1842 Missense Mutation CTT,TTT L603F NP_055045.2

View Full Product Details