Product Details

SNP ID
rs184993923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:408325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCACCGTTTTCCCCAGACCCCC[A/G]AAGCCGCTCCAAGAACCACATGTTG
Phenotype
MIM: 610336 MIM: 605217
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2CD4C PubMed Links

Gene Details

Gene
C2CD4C
Gene Name
C2 calcium dependent domain containing 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136263.1 200 Missense Mutation CGG,TGG R13W NP_001129735.1
XM_011527694.1 200 Missense Mutation CGG,TGG R13W XP_011525996.1
Gene
SHC2
Gene Name
SHC adaptor protein 2
There are no transcripts associated with this gene.

View Full Product Details