Product Details
- SNP ID
-
rs184771146
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:34111421 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGCATGACCCTGGCGTTGAAGCC[A/G]TGTCCTCCCAGGTAGCCGTTCCTGG
- Phenotype
-
MIM: 606745
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PARD3
PubMed Links
Gene Details
- Gene
- PARD3
- Gene Name
- par-3 family cell polarity regulator
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001184785.1 |
3972 |
Silent Mutation |
CAC,CAT |
H1270H |
NP_001171714.1 |
NM_001184786.1 |
3972 |
Silent Mutation |
CAC,CAT |
H1257H |
NP_001171715.1 |
NM_001184787.1 |
3972 |
Silent Mutation |
CAC,CAT |
H1236H |
NP_001171716.1 |
NM_001184788.1 |
3972 |
Silent Mutation |
CAC,CAT |
H1227H |
NP_001171717.1 |
NM_001184789.1 |
3972 |
Silent Mutation |
CAC,CAT |
H1190H |
NP_001171718.1 |
NM_001184790.1 |
3972 |
Silent Mutation |
CAC,CAT |
H1183H |
NP_001171719.1 |
NM_001184791.1 |
3972 |
Silent Mutation |
CAC,CAT |
H1161H |
NP_001171720.1 |
NM_001184792.1 |
3972 |
Intron |
|
|
NP_001171721.1 |
NM_001184793.1 |
3972 |
Intron |
|
|
NP_001171722.1 |
NM_001184794.1 |
3972 |
Intron |
|
|
NP_001171723.1 |
NM_019619.3 |
3972 |
Silent Mutation |
CAC,CAT |
H1273H |
NP_062565.2 |
XM_005252528.4 |
3972 |
Silent Mutation |
CAC,CAT |
H1269H |
XP_005252585.1 |
XM_005252530.4 |
3972 |
Silent Mutation |
CAC,CAT |
H1255H |
XP_005252587.1 |
XM_005252531.4 |
3972 |
Silent Mutation |
CAC,CAT |
H1240H |
XP_005252588.1 |
XM_005252532.4 |
3972 |
Silent Mutation |
CAC,CAT |
H1226H |
XP_005252589.1 |
XM_005252534.4 |
3972 |
Intron |
|
|
XP_005252591.1 |
XM_005252535.4 |
3972 |
Intron |
|
|
XP_005252592.1 |
XM_005252536.4 |
3972 |
Intron |
|
|
XP_005252593.1 |
XM_011519569.2 |
3972 |
Silent Mutation |
CAC,CAT |
H1272H |
XP_011517871.1 |
XM_011519570.2 |
3972 |
Silent Mutation |
CAC,CAT |
H1260H |
XP_011517872.1 |
XM_011519571.2 |
3972 |
Silent Mutation |
CAC,CAT |
H1258H |
XP_011517873.1 |
XM_011519572.2 |
3972 |
Silent Mutation |
CAC,CAT |
H1257H |
XP_011517874.1 |
XM_011519573.2 |
3972 |
Silent Mutation |
CAC,CAT |
H1243H |
XP_011517875.1 |
XM_011519575.2 |
3972 |
Silent Mutation |
CAC,CAT |
H1239H |
XP_011517877.1 |
XM_011519576.2 |
3972 |
Silent Mutation |
CAC,CAT |
H1229H |
XP_011517878.1 |
XM_011519577.2 |
3972 |
Silent Mutation |
CAC,CAT |
H1217H |
XP_011517879.1 |
XM_011519578.2 |
3972 |
Silent Mutation |
CAC,CAT |
H1215H |
XP_011517880.1 |
XM_011519579.2 |
3972 |
Silent Mutation |
CAC,CAT |
H1195H |
XP_011517881.1 |
XM_011519581.2 |
3972 |
Intron |
|
|
XP_011517883.1 |
XM_011519583.2 |
3972 |
Intron |
|
|
XP_011517885.1 |
XM_011519584.2 |
3972 |
Intron |
|
|
XP_011517886.1 |
XM_017016430.1 |
3972 |
Intron |
|
|
XP_016871919.1 |
View Full Product Details