Product Details

SNP ID
rs184581509
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:169921738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAATCCATATGCTGTGGCAGGGC[A/G]TCCAAGAATGCCAACTGGTTGGCCA
Phenotype
MIM: 601836
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIFAP3 PubMed Links

Gene Details

Gene
KIFAP3
Gene Name
kinesin associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204514.1 2752 Missense Mutation NP_001191443.1
NM_001204516.1 2752 Missense Mutation NP_001191445.1
NM_001204517.1 2752 Missense Mutation NP_001191446.1
NM_014970.3 2752 Missense Mutation NP_055785.2
XM_005244970.1 2752 Missense Mutation XP_005245027.1
XM_011509307.2 2752 UTR 3 XP_011507609.2

View Full Product Details