Product Details

SNP ID
rs184570913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49156253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGGTGAATTTGCACAGATCTGG[A/C]CAGATGCTACGAGGAACAAGATAGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C10orf128 PubMed Links

Gene Details

Gene
C10orf128
Gene Name
chromosome 10 open reading frame 128
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010863.2 521 Intron NP_001010863.1
NM_001288740.1 521 Intron NP_001275669.1
NM_001288741.1 521 Intron NP_001275670.1
NM_001288742.1 521 Missense Mutation GGC,GTC G90V NP_001275671.1
NM_001288743.1 521 Missense Mutation GCC,TCC A153S NP_001275672.1
XM_011539378.2 521 Intron XP_011537680.1
XM_011539379.1 521 Intron XP_011537681.1
XM_011539380.1 521 Intron XP_011537682.1
XM_011539382.1 521 Intron XP_011537684.1
XM_011539383.2 521 Missense Mutation TGG,TGT W100C XP_011537685.1
XM_011539384.1 521 Intron XP_011537686.1
XM_011539385.1 521 Intron XP_011537687.1
XM_017015792.1 521 Intron XP_016871281.1
XM_017015793.1 521 Intron XP_016871282.1
XM_017015794.1 521 Intron XP_016871283.1
XM_017015795.1 521 Intron XP_016871284.1
XM_017015796.1 521 Intron XP_016871285.1
XM_017015797.1 521 Intron XP_016871286.1
Gene
FAM170B-AS1
Gene Name
FAM170B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details