Product Details

SNP ID
rs185459656
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59364995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTAATGTCATCAGACATTTCTT[C/T]TGTGACATGCCCCAACTGTTAATCT
Phenotype
MIM: 615702
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR5AN1 PubMed Links

Gene Details

Gene
OR5AN1
Gene Name
olfactory receptor family 5 subfamily AN member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004729.1 537 Silent Mutation TTC,TTT F179F NP_001004729.1

View Full Product Details