Product Details

SNP ID
rs185264593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:27748253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATATTTACCTCTTCAACTCCTCTC[A/G]CAGTGATGGTGCTCAATACAGTAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF8L2 PubMed Links

Gene Details

Gene
DCAF8L2
Gene Name
DDB1 and CUL4 associated factor 8 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136533.1 1445 Missense Mutation NP_001130005.1

View Full Product Details