Product Details

SNP ID
rs185241384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50979797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGGAGGGTCTCACATTGCAGGC[A/G]TTTTTCTTGGAGTCGGGGATGCCAG
Phenotype
MIM: 604438
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLK7 PubMed Links

Gene Details

Gene
KLK7
Gene Name
kallikrein related peptidase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207053.1 869 Silent Mutation AAC,AAT N127N NP_001193982.1
NM_001243126.1 869 Silent Mutation AAC,AAT N192N NP_001230055.1
NM_005046.3 869 Silent Mutation AAC,AAT N199N NP_005037.1
NM_139277.2 869 Silent Mutation AAC,AAT N199N NP_644806.1

View Full Product Details