Product Details

SNP ID
rs185737637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55220668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCATCGCAGCGCTCGGAAATCT[C/T]CCTCAGGATGGCGTCCACTTCCGCG
Phenotype
MIM: 614086
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCIDAS PubMed Links

Gene Details

Gene
MCIDAS
Gene Name
multiciliate differentiation and DNA synthesis associated cell cycle protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190787.1 1033 Missense Mutation NP_001177716.1
XM_017009439.1 1033 Missense Mutation XP_016864928.1

View Full Product Details