Product Details

SNP ID
rs185700167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28319240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGTCCAGGCCCAACCGACAGACT[A/G]TGGGGGCTCCGGCAACCAGGCGCTG
Phenotype
MIM: 614394 MIM: 612912
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFT20 PubMed Links

Gene Details

Gene
IFT20
Gene Name
intraflagellar transport 20
There are no transcripts associated with this gene.

Gene
TMEM97
Gene Name
transmembrane protein 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014573.2 200 Missense Mutation ATG,GTG M1V NP_055388.2
XM_005257965.3 200 Silent Mutation CTA,CTG L41L XP_005258022.1

View Full Product Details