Product Details

SNP ID
rs186008754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143052881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCACTTCTTCTGTGACAATGAA[A/C]CTCTCCTGCAGTTGTCATGCTCTGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR6V1 PubMed Links

Gene Details

Gene
OR6V1
Gene Name
olfactory receptor family 6 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001667.1 541 Missense Mutation ACT,CCT T181P NP_001001667.1
Gene
OR6W1P
Gene Name
olfactory receptor family 6 subfamily W member 1 pseudogene
There are no transcripts associated with this gene.

View Full Product Details