Product Details

SNP ID
rs186107449
Assay Type
Functionally Tested
NCBI dbSNP Submissions
5
Location
Chr.11:55572179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCATTCTGCTTGGATTGACACAG[C/G]ATCCTTTTTGGAAGAAAATAGTGTT
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
OR4C16 PubMed Links

Gene Details

Gene
OR4C16
Gene Name
olfactory receptor family 4 subfamily C member 16 (gene/pseudogene)
There are no transcripts associated with this gene.

View Full Product Details