Product Details

SNP ID
rs186548772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70777656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTACCTGTGACTCAGCAGTTCTA[C/T]TTCTGGGAATCTATCCTGCAAAAAC
Phenotype
MIM: 102681 MIM: 608697
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADD2 PubMed Links

Gene Details

Gene
ADD2
Gene Name
adducin 2
There are no transcripts associated with this gene.

Gene
FIGLA
Gene Name
folliculogenesis specific bHLH transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004311.3 630 Missense Mutation ATA,GTA I209V NP_001004311.2

View Full Product Details