Product Details

SNP ID
rs186576711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102807424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGGTCCCAAAGGAGATGGTCAT[C/T]ATCCATGGAGTTGTCCAGTGACTCA
Phenotype
MIM: 611613
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BIVM PubMed Links

Gene Details

Gene
BIVM
Gene Name
basic, immunoglobulin-like variable motif containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159596.1 279 Intron NP_001153068.1
NM_017693.3 279 Missense Mutation CAT,TAT H53Y NP_060163.2
Gene
BIVM-ERCC5
Gene Name
BIVM-ERCC5 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204425.1 279 Missense Mutation CAT,TAT H53Y NP_001191354.1
Gene
KDELC1
Gene Name
KDEL motif containing 1
There are no transcripts associated with this gene.

View Full Product Details